Product Details

SNP ID
rs11880534
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:5563331 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAAGGGCAGGGGCTGCGGACCTTGG[C/T]GGGGGGCAACTGGGCTGGGCCAGGG
Phenotype
MIM: 615241
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TINCR PubMed Links

Gene Details

Gene
TINCR
Gene Name
tissue differentiation-inducing non-protein coding RNA
There are no transcripts associated with this gene.

View Full Product Details