Product Details
- SNP ID
-
rs12243034
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:71400322 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTGAAAGGTCATCGCATGGCTCAG[A/G]CATCCCACCCTGCCCAAAGAGGCAG
- Phenotype
-
MIM: 605516
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CDH23
PubMed Links
Gene Details
- Gene
- CDH23
- Gene Name
- cadherin-related 23
View Full Product Details