Product Details

SNP ID
rs11918599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:11734122 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTCCGGGGTGGGCAGCCACTTCCC[A/G]TCACCGGCTCTGTGCAAAGAGAACC
Phenotype
MIM: 614948
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAMM41 PubMed Links

Gene Details

Gene
TAMM41
Gene Name
TAM41 mitochondrial translocator assembly and maintenance homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284401.1 Intron NP_001271330.1
NM_001321294.1 Intron NP_001308223.1
NM_001321295.1 Intron NP_001308224.1
NM_138807.3 Intron NP_620162.1
XM_005264873.3 Intron XP_005264930.1
XM_005264875.3 Intron XP_005264932.1
XM_017005725.1 Intron XP_016861214.1
XM_017005726.1 Intron XP_016861215.1
XM_017005727.1 Intron XP_016861216.1
XM_017005728.1 Intron XP_016861217.1

View Full Product Details