Product Details

SNP ID
rs13264353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144879053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGCTCTTCTCCAAAGAAACAG[A/T]TAAGCTCTTGTCTTCTCTGAACTAA
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZNF250 PubMed Links

Gene Details

Gene
ZNF250
Gene Name
zinc finger protein 250
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109689.3 4262 UTR 3 NP_001103159.1
NM_021061.4 4262 UTR 3 NP_066405.1
XM_005272327.1 4262 Intron XP_005272384.1
XM_005272328.3 4262 Intron XP_005272385.1
XM_006716612.3 4262 UTR 3 XP_006716675.1
XM_006716613.3 4262 UTR 3 XP_006716676.1
XM_011517209.1 4262 Intron XP_011515511.1

View Full Product Details