Product Details

SNP ID
rs13270979
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66668462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTGATTTTTACGCTGCTGCTTACC[A/T]AAAAATCCCGGCCGGGCGCGGTGGC
Phenotype
MIM: 611745
Polymorphism
A/T, Transversion Substitution
Allele Nomenclature
Literature Links
C8orf44 PubMed Links
Additional Information
For this assay, SNP(s) [rs143635222] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
C8orf44
Gene Name
chromosome 8 open reading frame 44
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019607.2 Intron NP_062553.1
Gene
C8orf44-SGK3
Gene Name
C8orf44-SGK3 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001204173.1 Intron NP_001191102.1
Gene
VCPIP1
Gene Name
valosin containing protein interacting protein 1
There are no transcripts associated with this gene.

View Full Product Details