Product Details

SNP ID
rs12276274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:104887434 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGGAGAAGAGAGGGAAACCAATGCT[A/G]AATAAAGATGGACCTCCAACTTCCA
Phenotype
MIM: 608633
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CASP12 PubMed Links

Gene Details

Gene
CASP12
Gene Name
caspase 12 (gene/pseudogene)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001191016.2 Intron NP_001177945.2

View Full Product Details