Product Details

SNP ID
rs12225346
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:128698524 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTCTCACTTTATGGGTGGGGAAA[A/C]TGAGGTCTAGGTTGTTAAATGAATT
Phenotype
MIM: 193067 MIM: 615815
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FLI1 PubMed Links

Gene Details

Gene
FLI1
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001167681.2 Intron NP_001161153.1
NM_001271010.1 Intron NP_001257939.1
NM_001271012.1 Intron NP_001257941.1
NM_002017.4 Intron NP_002008.2
XM_011542701.2 Intron XP_011541003.1
XM_011542702.1 Intron XP_011541004.1
XM_017017405.1 Intron XP_016872894.1
XM_017017406.1 Intron XP_016872895.1
Gene
SENCR
Gene Name
smooth muscle and endothelial cell enriched migration/differentiation-associated long non-coding RNA
There are no transcripts associated with this gene.

View Full Product Details