Product Details
- SNP ID
-
rs11031586
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:32092833 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGAGAACACAGCCATCCCTGTG[C/T]TCCTGGGGGCCTCCAAAGGCCAAAG
- Phenotype
-
MIM: 602735
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RCN1
PubMed Links
Gene Details
- Gene
- RCN1
- Gene Name
- reticulocalbin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002901.2 |
|
Intron |
|
|
NP_002892.1 |
View Full Product Details