Product Details

SNP ID
rs10897078
Assay Type
Functionally Tested
NCBI dbSNP Submissions
35
Location
Chr.11:60518055 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAATTACATTATTTCGGTACTAACA[G/T]AATTCTCTTCTCACAGACTATCCAG
Phenotype
Polymorphism
G/T, Transversion Substitution
Allele Nomenclature
Literature Links
MS4A13 PubMed Links

Gene Details

Gene
MS4A13
Gene Name
membrane spanning 4-domains A13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012417.2 Intron NP_001012417.2
NM_001100909.1 Intron NP_001094379.1
NM_001278320.1 Intron NP_001265249.1

View Full Product Details