Product Details

SNP ID
rs11466049
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3243074 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGCAGCTAGCACCTAGTCGGCATTC[C/T]GTGACTATTGAGTGTGAATGCAAGA
Phenotype
MIM: 608107 MIM: 603231
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MEFV PubMed Links

Gene Details

Gene
MEFV
Gene Name
Mediterranean fever
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000243.2 2453 UTR 3 NP_000234.1
NM_001198536.1 2453 UTR 3 NP_001185465.1
XM_017023236.1 2453 Intron XP_016878725.1
Gene
ZNF200
Gene Name
zinc finger protein 200
There are no transcripts associated with this gene.

View Full Product Details