Product Details

SNP ID
rs4840666
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:7038561 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAAATTATGAACTCTTTTTCTCTA[G/T]TCTGCAAGCTTAGTAGCAGCAGATT
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DEFA11P PubMed Links

Gene Details

Gene
DEFA11P
Gene Name
defensin alpha 11, pseudogene
There are no transcripts associated with this gene.

Gene
DEFA7P
Gene Name
defensin alpha 7, pseudogene
There are no transcripts associated with this gene.

View Full Product Details