Product Details
- SNP ID
-
hCV32385067
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:49056720 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGACAAGTTATCTGAGGAAAAACA[C/A]CTGTTTCAAATGTATCAAGTGAGAC
- Phenotype
-
MIM: 300947
- Polymorphism
- C/A, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CCDC120
PubMed Links
Gene Details
- Gene
- CCDC120
- Gene Name
- coiled-coil domain containing 120
There are no transcripts associated with this gene.
View Full Product Details