Product Details
- SNP ID
-
rs16997824
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:26559915 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGTAAGGATAATGGAATCATTTTG[C/T]ATAAGTAAAAGAAAATGTCGTGTAG
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
VENTXP1
PubMed Links
Gene Details
- Gene
- VENTXP1
- Gene Name
- VENT homeobox pseudogene 1
There are no transcripts associated with this gene.
View Full Product Details