Product Details

SNP ID
rs34549705
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:32239394 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGCTGATCTCTGCCATGTCTGA[A/G]CCGGCCACTGTGGCAGCCCCTGTGC
Phenotype
MIM: 603911
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EIF3I PubMed Links

Gene Details

Gene
EIF3I
Gene Name
eukaryotic translation initiation factor 3 subunit I
There are no transcripts associated with this gene.

Gene
FAM167B
Gene Name
family with sequence similarity 167 member B
There are no transcripts associated with this gene.

Gene
MTMR9LP
Gene Name
myotubularin related protein 9-like, pseudogene
There are no transcripts associated with this gene.

View Full Product Details