Product Details
- SNP ID
-
rs11554833
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.19:49659723 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTAGGCCTTGTACTGGTCGGAGGT[A/G]AGGGAGAGTGGGTGGCTGTTGGAAA
- Phenotype
-
MIM: 610837
MIM: 603734
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
BCL2L12
PubMed Links
Gene Details
- Gene
- BCL2L12
- Gene Name
- BCL2 like 12
There are no transcripts associated with this gene.
- Gene
- IRF3
- Gene Name
- interferon regulatory factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001197122.1 |
1520 |
Missense Mutation |
CAC,TAC |
H409Y |
NP_001184051.1 |
NM_001197123.1 |
1520 |
Silent Mutation |
CTC,CTT |
L368L |
NP_001184052.1 |
NM_001197124.1 |
1520 |
Silent Mutation |
CTC,CTT |
L276L |
NP_001184053.1 |
NM_001197125.1 |
1520 |
Silent Mutation |
CTC,CTT |
L257L |
NP_001184054.1 |
NM_001197126.1 |
1520 |
Silent Mutation |
CTC,CTT |
L257L |
NP_001184055.1 |
NM_001197127.1 |
1520 |
Silent Mutation |
CTC,CTT |
L130L |
NP_001184056.1 |
NM_001197128.1 |
1520 |
Silent Mutation |
CTC,CTT |
L130L |
NP_001184057.1 |
NM_001571.5 |
1520 |
Silent Mutation |
CTC,CTT |
L403L |
NP_001562.1 |
XM_006723197.1 |
1520 |
Missense Mutation |
CAC,TAC |
H409Y |
XP_006723260.1 |
XM_006723198.1 |
1520 |
Missense Mutation |
CAC,TAC |
H409Y |
XP_006723261.1 |
XM_006723200.1 |
1520 |
Missense Mutation |
CAC,TAC |
H374Y |
XP_006723263.1 |
XM_006723201.1 |
1520 |
Missense Mutation |
CAC,TAC |
H263Y |
XP_006723264.1 |
XM_006723202.2 |
1520 |
Missense Mutation |
CAC,TAC |
H263Y |
XP_006723265.1 |
XM_017026766.1 |
1520 |
Silent Mutation |
CTC,CTT |
L403L |
XP_016882255.1 |
XM_017026767.1 |
1520 |
Silent Mutation |
CTC,CTT |
L403L |
XP_016882256.1 |
- Gene
- SCAF1
- Gene Name
- SR-related CTD associated factor 1
There are no transcripts associated with this gene.
View Full Product Details