Product Details

SNP ID
rs12946329
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:80221973 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GACCCAGGCTCCTATGCCTGTTTGG[C/T]ACACACAGACACTGAGCTGGTTATG
Phenotype
MIM: 605270 MIM: 610117
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGSH PubMed Links

Gene Details

Gene
SGSH
Gene Name
N-sulfoglucosamine sulfohydrolase
There are no transcripts associated with this gene.

Gene
SLC26A11
Gene Name
solute carrier family 26 member 11
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166347.1 Intron NP_001159819.1
NM_001166348.1 Intron NP_001159820.1
NM_001166349.1 Intron NP_001159821.1
NM_173626.3 Intron NP_775897.3
XM_006721833.1 Intron XP_006721896.1
XM_011524652.1 Intron XP_011522954.1
XM_011524653.1 Intron XP_011522955.1
XM_011524654.1 Intron XP_011522956.1
XM_017024505.1 Intron XP_016879994.1
XM_017024506.1 Intron XP_016879995.1
XM_017024507.1 Intron XP_016879996.1

View Full Product Details