Product Details
- SNP ID
-
rs6958331
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:55372830 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCCTTTTTTTCCTAAGGAAAATA[C/T]GGTTAGATGTACAATATTTATATGG
- Phenotype
-
MIM: 612919
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LANCL2
PubMed Links
Gene Details
- Gene
- LANCL2
- Gene Name
- LanC like 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018697.3 |
|
Intron |
|
|
NP_061167.1 |
View Full Product Details