Product Details

SNP ID
rs6868856
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:131159792 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATCTACAACTGGAGACTCATTAAG[A/G]TTAGGTATTTATCATTACTAATTTC
Phenotype
MIM: 601314
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HINT1 PubMed Links

Gene Details

Gene
HINT1
Gene Name
histidine triad nucleotide binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005340.6 Intron NP_005331.1
XM_017009410.1 Intron XP_016864899.1

View Full Product Details