Product Details

SNP ID
hCV57475963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.1:160211563 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGTCATGGCTGAGTACGGGACC[C/T]TCCTGCAAGACCTGACCAACAACAT
Phenotype
MIM: 114250 MIM: 615820 MIM: 603434
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CASQ1 PubMed Links

Gene Details

Gene
CASQ1
Gene Name
calsequestrin 1
There are no transcripts associated with this gene.

Gene
DCAF8
Gene Name
DDB1 and CUL4 associated factor 8
There are no transcripts associated with this gene.

Gene
LOC729867
Gene Name
uncharacterized LOC729867
There are no transcripts associated with this gene.

Gene
PEA15
Gene Name
phosphoprotein enriched in astrocytes 15
There are no transcripts associated with this gene.

View Full Product Details