Product Details

SNP ID
rs34851419
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:16044504 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCCAGGGGCCTGATGGAGGAGTT[G/T]GTGGGGCTGCGTGAAGGCTCCTCAG
Phenotype
MIM: 602023
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLCNKB PubMed Links

Gene Details

Gene
CLCNKB
Gene Name
chloride voltage-gated channel Kb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000085.4 164 Missense Mutation TTG,TTT L4F NP_000076.2
NM_001165945.2 164 Intron NP_001159417.2

View Full Product Details