Product Details
- SNP ID
-
rs28665609
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:108549155 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATAATGATGCACTTCCTAAGTTT[G/T]GTTTTTTCCATTAGGAAATGGACAT
- Phenotype
-
MIM: 610643
MIM: 609929
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KIAA1524
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76734463] are located under a probe and SNP(s) [rs114847492] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KIAA1524
- Gene Name
- KIAA1524
- Gene
- MYH15
- Gene Name
- myosin heavy chain 15
View Full Product Details