Product Details

SNP ID
rs28682868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:66224822 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAGCAGTTCTATTAATAGTGATT[C/T]CTCAGATACGTTTATTGCAGTGTTT
Phenotype
MIM: 603125
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TPST1 PubMed Links

Gene Details

Gene
TPST1
Gene Name
tyrosylprotein sulfotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003596.3 Intron NP_003587.1
XM_005250642.2 Intron XP_005250699.1
XM_011516634.2 Intron XP_011514936.1
XM_011516635.2 Intron XP_011514937.1
XM_017012724.1 Intron XP_016868213.1
XM_017012725.1 Intron XP_016868214.1
XM_017012726.1 Intron XP_016868215.1
XM_017012727.1 Intron XP_016868216.1

View Full Product Details