Product Details

SNP ID
rs28532769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:150791636 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCTTCCTCATTCTGGAAAAAAAA[A/G]AAAAAAGAAATCCTTAGGGGAAAAG
Phenotype
MIM: 610334 MIM: 610385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC105375566 PubMed Links

Gene Details

Gene
LOC105375566
Gene Name
uncharacterized LOC105375566
There are no transcripts associated with this gene.

Gene
TMEM176A
Gene Name
transmembrane protein 176A
There are no transcripts associated with this gene.

Gene
TMEM176B
Gene Name
transmembrane protein 176B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001101311.1 Intron NP_001094781.1
NM_001101312.1 Intron NP_001094782.1
NM_001101314.1 Intron NP_001094784.1
NM_014020.3 Intron NP_054739.3
XM_006715933.3 Intron XP_006715996.2

View Full Product Details