Product Details
- SNP ID
-
rs28641968
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:73423787 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGTTTACAGAAAACTTGGGCAGGTA[C/G]TACACAGAGTTCCCATATTCTTCCC
- Phenotype
-
MIM: 605920
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
STAU2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs75346547] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- STAU2
- Gene Name
- staufen double-stranded RNA binding protein 2
- Gene
- STAU2-AS1
- Gene Name
- STAU2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details