Product Details
- SNP ID
-
rs28547349
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.18:12957749 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTCCTGCTATTACCCCTCACCTGCC[A/C]TCCCTAATGGCATGATACTACGTTA
- Phenotype
-
MIM: 609263
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
SEH1L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs76454292] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- SEH1L
- Gene Name
- SEH1 like nucleoporin
View Full Product Details