Product Details

SNP ID
rs28627435
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:15736906 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATAATGCTTTTTGATGGTTTGATG[C/T]CACTCTGCTTTTTTTTTTTTTCCAG
Phenotype
MIM: 604736
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
USP25 PubMed Links

Gene Details

Gene
USP25
Gene Name
ubiquitin specific peptidase 25
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001283041.1 Intron NP_001269970.1
NM_001283042.1 Intron NP_001269971.1
NM_013396.4 Intron NP_037528.3
XM_005260949.4 Intron XP_005261006.1
XM_006723992.3 Intron XP_006724055.1
XM_011529529.1 Intron XP_011527831.1
XM_011529531.2 Intron XP_011527833.1
XM_011529533.2 Intron XP_011527835.1
XM_011529535.2 Intron XP_011527837.1
XM_017028321.1 Intron XP_016883810.1
XM_017028322.1 Intron XP_016883811.1
XM_017028323.1 Intron XP_016883812.1
XM_017028324.1 Intron XP_016883813.1
XM_017028325.1 Intron XP_016883814.1
XM_017028327.1 Intron XP_016883816.1
XM_017028328.1 Intron XP_016883817.1
XM_017028329.1 Intron XP_016883818.1

View Full Product Details