Product Details

SNP ID
rs17466177
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:222679130 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAACATATATCAATGTACTGCCTTA[C/T]ATAATATGCACGAAAGAAGATAAAA
Phenotype
MIM: 612375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AIDA PubMed Links

Gene Details

Gene
AIDA
Gene Name
axin interactor, dorsalization associated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022831.2 Intron NP_073742.2

View Full Product Details