Product Details

SNP ID
rs17163445
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:222719612 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
ACTACAGTTGAACAGCCTTGACCTT[G/T]AATAACACACAAAGGATGATTTGAG
Phenotype
MIM: 612375
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
AIDA PubMed Links

Gene Details

Gene
AIDA
Gene Name
axin interactor, dorsalization associated
There are no transcripts associated with this gene.

Gene
BROX
Gene Name
BRO1 domain and CAAX motif containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288579.1 Intron NP_001275508.1
NM_001288580.1 Intron NP_001275509.1
NM_001288581.1 Intron NP_001275510.1
NM_144695.3 Intron NP_653296.2
XM_005273065.2 Intron XP_005273122.2
XM_005273069.4 Intron XP_005273126.1
XM_006711173.3 Intron XP_006711236.1
XM_011509212.2 Intron XP_011507514.1
XM_011509213.2 Intron XP_011507515.1
XM_011509214.2 Intron XP_011507516.1
XM_017000374.1 Intron XP_016855863.1
XM_017000375.1 Intron XP_016855864.1
XM_017000376.1 Intron XP_016855865.1

View Full Product Details