Product Details

SNP ID
rs17163453
Assay Type
Functionally tested
NCBI dbSNP Submissions
47
Location
Chr.1:222724322 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTAAGTATGTTATTGATGAGTGC[A/G]GCCAGTTACTAAATATTAAATGTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BROX PubMed Links

Gene Details

Gene
BROX
Gene Name
BRO1 domain and CAAX motif containing
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288579.1 Intron NP_001275508.1
NM_001288580.1 Intron NP_001275509.1
NM_001288581.1 Intron NP_001275510.1
NM_144695.3 Intron NP_653296.2
XM_005273065.2 Intron XP_005273122.2
XM_005273069.4 Intron XP_005273126.1
XM_006711173.3 Intron XP_006711236.1
XM_011509212.2 Intron XP_011507514.1
XM_011509213.2 Intron XP_011507515.1
XM_011509214.2 Intron XP_011507516.1
XM_017000374.1 Intron XP_016855863.1
XM_017000375.1 Intron XP_016855864.1
XM_017000376.1 Intron XP_016855865.1

View Full Product Details