Product Details

SNP ID
rs28786533
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:13972809 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGAGAATCCCAGATGGGCCCCCAA[A/G]TTTGCTAGAAATAAATTTTTGGTGC
Phenotype
MIM: 611167
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TPRXL PubMed Links

Gene Details

Gene
TPRXL
Gene Name
tetrapeptide repeat homeobox like
There are no transcripts associated with this gene.

View Full Product Details