Product Details

SNP ID
rs11551632
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:52949543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGGAGAAGAAGGGACCCCAGGTC[A/G]GAGACTGGAGCCATTACTTCAAGAT
Phenotype
MIM: 148070 MIM: 148060
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KRT18 PubMed Links

Gene Details

Gene
KRT18
Gene Name
keratin 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000224.2 Intron NP_000215.1
NM_199187.1 Intron NP_954657.1
Gene
KRT8
Gene Name
keratin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256282.1 Intron NP_001243211.1
NM_001256293.1 Intron NP_001243222.1
NM_002273.3 Intron NP_002264.1

View Full Product Details