Product Details

SNP ID
rs3207600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:64230922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCAACCCTCCCTCCCGGCTCCTCG[A/G]AGCGGCCGCCGGGCAGCGGTGAGTT
Phenotype
MIM: 604189 MIM: 607901 MIM: 610470 MIM: 601398
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNAJC4 PubMed Links

Gene Details

Gene
DNAJC4
Gene Name
DnaJ heat shock protein family (Hsp40) member C4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307980.1 491 Missense Mutation GAA,GGA E23G NP_001294909.1
NM_001307981.1 491 Missense Mutation GAA,GGA E23G NP_001294910.1
NM_005528.3 491 Missense Mutation GAA,GGA E23G NP_005519.2
Gene
FERMT3
Gene Name
fermitin family member 3
There are no transcripts associated with this gene.

Gene
NUDT22
Gene Name
nudix hydrolase 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001128612.2 491 Intron NP_001122084.1
NM_001128613.2 491 Intron NP_001122085.1
NM_001271831.1 491 Intron NP_001258760.1
NM_032344.3 491 Intron NP_115720.1
Gene
TRPT1
Gene Name
tRNA phosphotransferase 1
There are no transcripts associated with this gene.

Gene
VEGFB
Gene Name
vascular endothelial growth factor B
There are no transcripts associated with this gene.

View Full Product Details