Product Details

SNP ID
rs61742093
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:27912204 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGAATTATTGTCAGATTGCCAAAG[A/G]TTGTCAAGATATAGGAAAACAGAAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR2B2 PubMed Links

Gene Details

Gene
OR2B2
Gene Name
olfactory receptor family 2 subfamily B member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033057.2 193 Missense Mutation ACC,ATC T39I NP_149046.2

View Full Product Details