Product Details
- SNP ID
-
rs61734927
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:156950316 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGATCCGAAAGCTGGAGCGTTGCCT[A/C]GGTTTGAACAAGCGCAAAAAGAAGG
- Phenotype
-
MIM: 611269
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NOM1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs142959370] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NOM1
- Gene Name
- nucleolar protein with MIF4G domain 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138400.1 |
594 |
Silent Mutation |
CTA,CTC |
L193L |
NP_612409.1 |
View Full Product Details