Product Details

SNP ID
rs60978388
Assay Type
Functionally tested
NCBI dbSNP Submissions
14
Location
Chr.1:165724610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGGGGCCCAGCAGTTTGTGTT[C/T]TAACAAGGCTCTAGTGATTCTGATA
Phenotype
MIM: 614123
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TMCO1 PubMed Links

Gene Details

Gene
TMCO1
Gene Name
transmembrane and coiled-coil domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256164.1 4351 UTR 3 NP_001243093.1
NM_001256165.1 4351 UTR 3 NP_001243094.1
NM_019026.4 4351 UTR 3 NP_061899.2

View Full Product Details