Product Details

SNP ID
rs61621288
Assay Type
Functionally tested
NCBI dbSNP Submissions
19
Location
Chr.1:165726272 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCCTTCTGATAAAACTAGTCTCA[A/G]AATAGGGACTTCTGTTCCCTTTCAT
Phenotype
MIM: 614123
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMCO1 PubMed Links

Gene Details

Gene
TMCO1
Gene Name
transmembrane and coiled-coil domains 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256164.1 2689 UTR 3 NP_001243093.1
NM_001256165.1 2689 UTR 3 NP_001243094.1
NM_019026.4 2689 UTR 3 NP_061899.2

View Full Product Details