Product Details

SNP ID
rs56184971
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:134782343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTTGGTGGGTAGATTTTTAATTA[C/T]TGATTCAAATTCTTTTGTGAAAATC
Phenotype
MIM: 600600
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EPHB1 PubMed Links

Gene Details

Gene
EPHB1
Gene Name
EPH receptor B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004441.4 Intron NP_004432.1
XM_011512542.1 Intron XP_011510844.1
XM_017005866.1 Intron XP_016861355.1
XM_017005867.1 Intron XP_016861356.1
XM_017005868.1 Intron XP_016861357.1

View Full Product Details