Product Details

SNP ID
rs61000990
Assay Type
Functionally tested
NCBI dbSNP Submissions
33
Location
Chr.1:114587732 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAACTGACCTTTAACTCCACTTTT[C/T]CGAAGCTCTCGGTCCTGGATGAATC
Phenotype
MIM: 605783
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
BCAS2 PubMed Links

Gene Details

Gene
BCAS2
Gene Name
breast carcinoma amplified sequence 2
There are no transcripts associated with this gene.

Gene
DENND2C
Gene Name
DENN domain containing 2C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256404.1 3354 Silent Mutation CGA,CGG R884R NP_001243333.1
NM_198459.3 3354 Silent Mutation CGA,CGG R827R NP_940861.3

View Full Product Details