Product Details

SNP ID
rs56350877
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:127307759 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCATTGCTTCAAATTCAGCCCA[A/G]GGCGGCTTTTCAGTTAGCATTTCTA
Phenotype
MIM: 609487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MAP3K2 PubMed Links

Gene Details

Gene
MAP3K2
Gene Name
mitogen-activated protein kinase kinase kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006609.4 1780 Silent Mutation CCC,CCT P560P NP_006600.3

View Full Product Details