Product Details
- SNP ID
-
hCV90397322
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:86960113 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCGGAGAACATCGCGGTCACCTC[A/C]GGGGTGGTGCGCAAGGTGAGCCCCG
- Phenotype
-
MIM: 608925
MIM: 602998
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ADIRF
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs111800178] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADIRF
- Gene Name
- adipogenesis regulatory factor
There are no transcripts associated with this gene.
- Gene
- MMRN2
- Gene Name
- multimerin 2
There are no transcripts associated with this gene.
- Gene
- SNCG
- Gene Name
- synuclein gamma
There are no transcripts associated with this gene.
View Full Product Details