Product Details

SNP ID
rs73138112
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:78485283 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGGGGAGGGGAGCACATAGTGAA[A/C]ATACATATGCACAGGAACCAAATCA
Phenotype
MIM: 609718 MIM: 606911
Polymorphism
A/C, Transversion Substitution
Allele Nomenclature
Literature Links
LHFPL2 PubMed Links

Gene Details

Gene
LHFPL2
Gene Name
lipoma HMGIC fusion partner-like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005779.2 4977 UTR 3 NP_005770.1
XM_006714515.1 4977 UTR 3 XP_006714578.1
Gene
SCAMP1
Gene Name
secretory carrier membrane protein 1
There are no transcripts associated with this gene.

View Full Product Details