Product Details

SNP ID
rs73614656
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149973648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTGTATAAAAATTTGTATGATGA[G/T]AATTGTATAATAATTATACATGAAA
Phenotype
MIM: 605697
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ULBP1 PubMed Links

Gene Details

Gene
ULBP1
Gene Name
UL16 binding protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317089.1 2981 Intron NP_001304018.1
NM_025218.3 2981 Intron NP_079494.1
XM_017011322.1 2981 UTR 3 XP_016866811.1

View Full Product Details