Product Details

SNP ID
rs74061089
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73276096 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGTTGCTGATATGCCCTCCTGGG[A/G]ATTCTGGCAGCATAGCGTGCCTCTC
Phenotype
MIM: 603728
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NUMB PubMed Links

Gene Details

Gene
NUMB
Gene Name
NUMB, endocytic adaptor protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001005743.1 2725 UTR 3 NP_001005743.1
NM_001005744.1 2725 UTR 3 NP_001005744.1
NM_001005745.1 2725 UTR 3 NP_001005745.1
NM_001320114.1 2725 UTR 3 NP_001307043.1
NM_003744.5 2725 UTR 3 NP_003735.3
Gene
PAPLN
Gene Name
papilin, proteoglycan like sulfated glycoprotein
There are no transcripts associated with this gene.

View Full Product Details