Product Details

SNP ID
rs72800312
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134655957 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGTGTTCTTCAAGGTGACCTAT[G/T]TTTCTTACTCTTATATACTCCCTGG
Phenotype
MIM: 607183
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SEC24A PubMed Links

Gene Details

Gene
SEC24A
Gene Name
SEC24 homolog A, COPII coat complex component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252231.1 Intron NP_001239160.1
NM_021982.2 Intron NP_068817.1
XM_006714523.2 Intron XP_006714586.1
XM_017008961.1 Intron XP_016864450.1
XM_017008962.1 Intron XP_016864451.1
XM_017008963.1 Intron XP_016864452.1

View Full Product Details