Product Details
- SNP ID
-
rs73790341
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:138766800 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTAAGTGCAGGGAGAAGCCCTATAC[G/T]TGTGAAAATGCAGCTCTGGTGAAGG
- Phenotype
-
MIM: 116805
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CTNNA1
PubMed Links
Gene Details
- Gene
- CTNNA1
- Gene Name
- catenin alpha 1
View Full Product Details