Product Details

SNP ID
rs78235896
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:160741794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAGTCTGTGAAATCCTGCCTTAG[A/G]AGCCTTGGCTCCTAATCCTGGCCTT
Phenotype
MIM: 606625
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLAMF7 PubMed Links

Gene Details

Gene
SLAMF7
Gene Name
SLAM family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282588.1 Intron NP_001269517.1
NM_001282589.1 Intron NP_001269518.1
NM_001282590.1 Intron NP_001269519.1
NM_001282591.1 Intron NP_001269520.1
NM_001282592.1 Intron NP_001269521.1
NM_001282593.1 Intron NP_001269522.1
NM_001282594.1 Intron NP_001269523.1
NM_001282595.1 Intron NP_001269524.1
NM_001282596.1 Intron NP_001269525.1
NM_021181.4 Intron NP_067004.3
XM_011509828.1 Intron XP_011508130.1
XM_011509829.1 Intron XP_011508131.1

View Full Product Details