Product Details

SNP ID
rs27829
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11255546 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGTGCGCTGCGGGAGAGACAAAG[A/C]GGTGAGCTGGGGCGCTGCGGGGCCG
Phenotype
MIM: 603597
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SOCS1 PubMed Links

Gene Details

Gene
SOCS1
Gene Name
suppressor of cytokine signaling 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003745.1 Intron NP_003736.1

View Full Product Details