Product Details

SNP ID
rs727030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:223878481 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAACCACTATGGACAGACCTTT[C/T]CCATATTAGTCCCCATGGGTAAGTT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WDFY1 PubMed Links

Gene Details

Gene
WDFY1
Gene Name
WD repeat and FYVE domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020830.4 1526 UTR 3 NP_065881.1

View Full Product Details