Product Details

SNP ID
rs175442
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:75136861 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTGAGAAAGAAATTAACTGTCTTC[C/T]GCTCTCAAGTTAGACCCCTAAGAAA
Phenotype
MIM: 609798 MIM: 605406
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NEK9 PubMed Links

Gene Details

Gene
NEK9
Gene Name
NIMA related kinase 9
There are no transcripts associated with this gene.

Gene
TMED10
Gene Name
transmembrane p24 trafficking protein 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006827.5 Intron NP_006818.3

View Full Product Details