Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_018717.4 | 4348 | UTR 3 | NP_061187.2 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001204366.1 | 4348 | Intron | NP_001191295.1 | ||
NM_001204367.1 | 4348 | Intron | NP_001191296.1 | ||
NM_001204368.1 | 4348 | Intron | NP_001191297.1 | ||
NM_002413.4 | 4348 | Intron | NP_002404.1 | ||
XM_006714221.2 | 4348 | Intron | XP_006714284.1 | ||
XM_011531972.2 | 4348 | Intron | XP_011530274.1 | ||
XM_017008211.1 | 4348 | Intron | XP_016863700.1 | ||
XM_017008212.1 | 4348 | Intron | XP_016863701.1 | ||
XM_017008213.1 | 4348 | Intron | XP_016863702.1 | ||
XM_017008214.1 | 4348 | Intron | XP_016863703.1 |